Ethyl-5-bromo-1H-pyrrolo[2,3-c]pyridine-2-carboxylate ≥99%
Supplier: Matrix Scientific
Synonyms:
Ethyl 5-bromo-1H-pyrrolo-[2,3-c]-pyridine-2-carboxylate
057526-1G
101913-228EA
869.23
USD
101913-228
Ethyl-5-bromo-1H-pyrrolo[2,3-c]pyridine-2-carboxylate ≥99%
Ethyl-5-bromo-1H-pyrrolo[2,3-c]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉BrN₂O₂ MW: 269.1 g/mol |
MDL Number:
MFCD12031217 CAS Number: 800401-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...