4-Chloro-6,7-dimethoxyquinoline ≥95%

Supplier: Matrix Scientific

057492-5G
101913-220EA 46 USD
101913-220
4-Chloro-6,7-dimethoxyquinoline ≥95%
4-Chloro-6,7-dimethoxyquinoline

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₂
MW: 223.6556 g/mol
MDL Number: MFCD07778437
CAS Number: 35654-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR