6-Methoxyquinolin-8-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Amino-6-methoxyquinoline

057491-1G
101913-216EA 769.23 USD
101913-216
6-Methoxyquinolin-8-amine ≥95%
6-Methoxyquinolin-8-amine

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
MDL Number: MFCD00672902
CAS Number: 90-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR