3-(Hydroxymethyl)-1-methylquinolin-2(1H)-one ≥95%

Supplier: Matrix Scientific

057489-1G
101913-212EA 613.85 USD
101913-212
3-(Hydroxymethyl)-1-methylquinolin-2(1H)-one ≥95%
3-(Hydroxymethyl)-1-methylquinolin-2(1H)-one

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD06738250
CAS Number: 114561-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR