8-Bromoquinoline-5-carboxylic acid ≥99%

Supplier: Matrix Scientific

057485-1G
101913-204EA 500 USD
101913-204
8-Bromoquinoline-5-carboxylic acid ≥99%
8-Bromoquinoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆BrNO₂
MW: 252.0600 g/mol
MDL Number: MFCD12963555
CAS Number: 204782-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR