8-Bromoquinoline-4-carbaldehyde ≥98%

Supplier: Matrix Scientific

057483-1G
101913-200EA 558.46 USD
101913-200
8-Bromoquinoline-4-carbaldehyde ≥98%
8-Bromoquinoline-4-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₆BrNO
MDL Number: MFCD06824368
CAS Number: 898391-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR