2-Aminothiazole-4-carboxylic acid hydrobromide ≥95%

Supplier: Matrix Scientific

057471-100G
101913-190EA 756.92 USD
101913-190
2-Aminothiazole-4-carboxylic acid hydrobromide ≥95%
2-Aminothiazole-4-carboxylic acid hydrobromide

Hazard: Irritant

Formula: C₄H₅BrN₂O₂S
MDL Number: MFCD02677963
CAS Number: 112539-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR