2-Chlorothiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

057470-1G
101913-186EA 73.85 USD
101913-186
2-Chlorothiazole-4-carboxylic acid ≥95%
2-Chlorothiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₄H₂ClNO₂S
MW: 163.58 g/mol
MDL Number: MFCD09870033
CAS Number: 5198-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR