tert-Butyl-4-formylbenzo[d]thiazol-2-ylcarbamate ≥98%

Supplier: Matrix Scientific

057467-1G
101913-180EA 613.85 USD
101913-180
tert-Butyl-4-formylbenzo[d]thiazol-2-ylcarbamate ≥98%
tert-Butyl-4-formylbenzo[d]thiazol-2-ylcarbamate

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₃S
MW: 278.33 g/mol
MDL Number: MFCD15072118
CAS Number: 1223748-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR