Ethyl-2-bromo-4-oxazolecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromooxazole-4-carboxylate

057455-1G
101913-162EA 115 USD
101913-162
Ethyl-2-bromo-4-oxazolecarboxylate ≥95%
Ethyl 2-bromo-4-oxazolecarboxylate

Hazard: Irritant/Store in Freezer

Formula: C₆H₆BrNO₃
MW: 220.0207 g/mol
MDL Number: MFCD09801023
CAS Number: 460081-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR