Cyclobutyl methyl ketone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Cyclobutylethan-1-one

057428-1G
101913-142EA 126.15 USD
101913-142
Cyclobutyl methyl ketone ≥95%
Cyclobutyl methyl ketone

Hazard: Flammable

Formula: C₆H₁₀O
MW: 98.15 g/mol
Boiling Pt: 134…135 °C
Density: 0.9
Flash Pt: 28 °C
MDL Number: MFCD00037148
CAS Number: 3019-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR