3-Oxetanone ≥97%
Supplier: Matrix Scientific
Synonyms:
Oxetan-3-one
057422-5G
101913-134EA
112.31
USD
101913-134
3-Oxetanone ≥97%
3-Oxetanone
Hazard: Irritant/Store in Freezer
Formula:
C₃H₄O₂ MW: 72.06 g/mol |
MDL Number:
MFCD09263255 CAS Number: 6704-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...