5-Bromo-2,4-dichloropyridine ≥95%

Supplier: Matrix Scientific

057406-25G
101913-110EA 1043.08 USD
101913-110
5-Bromo-2,4-dichloropyridine ≥95%
5-Bromo-2,4-dichloropyridine

Hazard: Irritant

Formula: C₅H₂BrCl₂N
MDL Number: MFCD09031035
CAS Number: 849937-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR