4-Methyl-6-trifluoromethylpyridin-3-ylamine ≥95%

Supplier: Matrix Scientific

057393-1G
101913-102EA 853.85 USD
101913-102
4-Methyl-6-trifluoromethylpyridin-3-ylamine ≥95%
4-Methyl-6-trifluoromethylpyridin-3-ylamine

Hazard: Irritant/Refrigerate

Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
MDL Number: MFCD11848206
CAS Number: 944317-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR