4-Bromo-2,6-diaminopyridine ≥95%

Supplier: Matrix Scientific

057391-1G
101913-096EA 313.85 USD
101913-096
4-Bromo-2,6-diaminopyridine ≥95%
4-Bromo-2,6-diaminopyridine

Hazard: Irritant

Formula: C₅H₆BrN₃
MDL Number: MFCD04115308
CAS Number: 329974-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR