3-Amino-6-chloro-4-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-4-methylpyridin-3-amine5-Amino-2-chloro-4-methylpyridine3-Amino-6-chloro-4-picoline

057383-25G
101913-084EA 92 USD
101913-084
3-Amino-6-chloro-4-methylpyridine ≥95%
3-Amino-6-chloro-4-methylpyridine

Hazard: Irritant

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD03427661
CAS Number: 66909-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR