5-Ethynyl-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

057364-250MG
101913-050EA 164.62 USD
101913-050
5-Ethynyl-2-methoxypyridine ≥95%
5-Ethynyl-2-methoxypyridine

Hazard: Irritant/Refrigerate

Formula: C₈H₇NO
MDL Number: MFCD10698135
CAS Number: 663955-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR