3,5-Dibenzo(d)oxazol-2-ylpyridine ≥95%

Supplier: Matrix Scientific

057351-1G
101913-028EA 490.77 USD
101913-028
3,5-Dibenzo(d)oxazol-2-ylpyridine ≥95%
3,5-Dibenzo(d)oxazol-2-ylpyridine

Hazard: Irritant

Formula: C₁₉H₁₁N₃O₂
MDL Number: MFCD15072117
CAS Number: 1223748-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR