2,4-Dichloro-6-methoxyquinazoline ≥95%

Supplier: Matrix Scientific

057335-1G
101913-000EA 976.35 USD
101913-000
2,4-Dichloro-6-methoxyquinazoline ≥95%
2,4-Dichloro-6-methoxyquinazoline

Hazard: Irritant

Formula: C₉H₆Cl₂N₂O
MW: 229.0627 g/mol
MDL Number: MFCD09954880
CAS Number: 105763-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR