2,4-Dichloro-7-fluoroquinazoline ≥99%

Supplier: Matrix Scientific

057328-1G
101912-986EA 384.1 USD
101912-986
2,4-Dichloro-7-fluoroquinazoline ≥99%
2,4-Dichloro-7-fluoroquinazoline

Hazard: Irritant

Formula: C₈H₃Cl₂FN₂
MW: 217.0272 g/mol
MDL Number: MFCD09954885
CAS Number: 174566-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR