6-Fluoroquinazolin-4-amine ≥98%

Supplier: Matrix Scientific

057318-1G
101912-978EA 846.15 USD
101912-978
6-Fluoroquinazolin-4-amine ≥98%
6-Fluoroquinazolin-4-amine

Hazard: Irritant

Formula: C₈H₆FN₃
MDL Number: MFCD12024574
CAS Number: 1190320-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR