(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: (3R,4R)-tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylatetrans-3-Amino-4-hydroxypyrrolidine, N1-BOC protected

057312-1G
101912-966EA 320 USD
101912-966
(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate ≥95%
(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₈N₂O₃
MW: 202.26 g/mol
MDL Number: MFCD11111251
CAS Number: 330681-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR