3-Methoxypyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

057309-1G
101912-960EA 146.15 USD
101912-960
3-Methoxypyrrolidine hydrochloride ≥95%
3-Methoxypyrrolidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MDL Number: MFCD09261403
CAS Number: 136725-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR