4,4-Dimethylpyrrolidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

057305-1G
101912-946EA 1503.08 USD
101912-946
4,4-Dimethylpyrrolidine-3-carboxylic acid ≥95%
4,4-Dimethylpyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₃NO₂
MDL Number: MFCD05662927
CAS Number: 261896-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR