tert-Butyl-3-(4-(aminomethyl)phenyl)pyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

057303-1G
101912-942EA 2850.77 USD
101912-942
tert-Butyl-3-(4-(aminomethyl)phenyl)pyrrolidine-1-carboxylate ≥95%
tert-Butyl-3-(4-(aminomethyl)phenyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₄N₂O₂
MW: 276.38 g/mol
MDL Number: MFCD08234643
CAS Number: 885270-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR