N-Cbz-4-oxo-D-proline ≥95%

Supplier: Matrix Scientific

057297-1G
101912-930EA 1306.15 USD
101912-930
N-Cbz-4-oxo-D-proline ≥95%
N-Cbz-4-oxo-D-proline

Hazard: Irritant

Formula: C₁₃H₁₃NO₅
MDL Number: MFCD09835491
CAS Number: 147226-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR