(Z)-Ethyl-3-(pyrrolidin-1-yl)but-2-enoate ≥95%

Supplier: Matrix Scientific

Synonyms: (Z)-Ethyl 3-(pyrrolidin-1-yl)but-2-enoate

057294-1G
101912-924EA 446.15 USD
101912-924
(Z)-Ethyl-3-(pyrrolidin-1-yl)but-2-enoate ≥95%
(Z)-Ethyl-3-(pyrrolidin-1-yl)but-2-enoate

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₇NO₂
MW: 183.25 g/mol
MDL Number: MFCD02081922
CAS Number: 70526-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR