(Octahydrocyclopenta[c]pyrrol-3a-yl)methanol ≥95%

Supplier: Matrix Scientific

057287-1G
101912-902EA 961.54 USD
101912-902
(Octahydrocyclopenta[c]pyrrol-3a-yl)methanol ≥95%
(Octahydrocyclopenta[c]pyrrol-3a-yl)methanol

Hazard: Irritant

Formula: C₈H₁₅NO
MDL Number: MFCD14708212
CAS Number: 444193-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR