(2S,4S)-1-Benzyl-2-methy-4-fluoropyrrolidine-1,2-dicarboxylate ≥91%

Supplier: Matrix Scientific

057270-1G
101912-870EA 1535.38 USD
101912-870
(2S,4S)-1-Benzyl-2-methy-4-fluoropyrrolidine-1,2-dicarboxylate ≥91%
(2S,4S)-1-Benzyl-2-methy-4-fluoropyrrolidine-1,2-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₁₆FNO₄
MDL Number: MFCD12963560
CAS Number: 72180-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR