3-(Piperidin-4-yl)benzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3-Cyanophenyl)piperidine

057236-1G
101912-810EA 370.77 USD
101912-810
3-(Piperidin-4-yl)benzonitrile ≥95%
3-(Piperidin-4-yl)benzonitrile

Hazard: Irritant

Formula: C₁₂H₁₄N₂
MW: 186.26 g/mol
MDL Number: MFCD03839908
CAS Number: 370864-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR