8-(Nitromethyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: Matrix Scientific

057223-1G
101912-788EA 286.15 USD
101912-788
8-(Nitromethyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(Nitromethyl)-1,4-dioxaspiro[4.5]decan-8-ol

Hazard: Irritant

Formula: C₉H₁₅NO₅
MDL Number: MFCD12828250
CAS Number: 879514-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR