3-Iodo-2-methylbenzhydrazide

Supplier: Matrix Scientific

057099-10G
101912-368EA 229.23 USD
101912-368
3-Iodo-2-methylbenzhydrazide
3-Iodo-2-methylbenzhydrazide

Hazard: Irritant

Formula: C₈H₉IN₂O
MDL Number: MFCD03425689
CAS Number: 1023473-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR