4-Fluorobenzyl thiocyanate ≥98%

Supplier: Matrix Scientific

057083-50G
101912-306EA 496.92 USD
101912-306
4-Fluorobenzyl thiocyanate ≥98%
4-Fluorobenzyl thiocyanate

Hazard: Irritant

Formula: C₈H₆FNS
MDL Number: MFCD09025694
CAS Number: 2740-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR