3-Fluorobenzyl thiocyanate ≥98%

Supplier: Matrix Scientific

057082-50G
101912-302EA 564.62 USD
101912-302
3-Fluorobenzyl thiocyanate ≥98%
3-Fluorobenzyl thiocyanate

Hazard: Irritant

Formula: C₈H₆FNS
MDL Number: MFCD09025693
CAS Number: 1174906-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR