4-(1-Pyrrolyl)benzonitrile

Supplier: Matrix Scientific

056988-5G 056988-1G
101911-978EA 470.77 USD
101911-978 101911-976
4-(1-Pyrrolyl)benzonitrile
4-(1-Pyrrolyl)benzonitrile

Hazard: Irritant

Formula: C₁₁H₈N₂
MDL Number: MFCD00085164
CAS Number: 23351-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR