2-Chloro-4-nitrothiobenzamide ≥98%

Supplier: Matrix Scientific

056976-2G 056976-10G
101911-930EA 535.38 USD
101911-930 101911-928
2-Chloro-4-nitrothiobenzamide ≥98%
2-Chloro-4-nitrothiobenzamide

Hazard: Irritant

Formula: C₇H₅CIN₂O₂S
MDL Number: MFCD09025682
CAS Number: 1233509-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR