5-Chloro-2-methylthiobenzamide ≥98%

Supplier: Matrix Scientific

056973-2G 056973-10G
101911-916EA 143.08 USD
101911-916 101911-918
5-Chloro-2-methylthiobenzamide ≥98%
5-Chloro-2-methylthiobenzamide

Hazard: Irritant

Formula: C₈H₈ClNS
MDL Number: MFCD09025681
CAS Number: 1174906-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR