3-Chloro-4-methylbenzhydrazide

Supplier: Matrix Scientific

056963-2G 056963-10G
101911-890EA 78.46 USD
101911-890 101911-892
3-Chloro-4-methylbenzhydrazide
3-Chloro-4-methylbenzhydrazide

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD03215573
CAS Number: 72198-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR