2-Iodothiobenzamide ≥98%

Supplier: Matrix Scientific

056853-1G
101911-540EA 55.38 USD
101911-540
2-Iodothiobenzamide ≥98%
2-Iodothiobenzamide

Hazard: Irritant

Formula: C₇H₆INS
MDL Number: MFCD09025699
CAS Number: 81568-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR