Dehydroabietyl isothiocyanate, Technical Grade

Supplier: Matrix Scientific

056699-1G
101911-226EA 126.15 USD
101911-226
Dehydroabietyl isothiocyanate, Technical Grade
Dehydroabietyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₂₁H₂₉NS
MDL Number: MFCD09025684
CAS Number: 115269-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR