1,3-Difluoroacetone ≥98%

Supplier: Matrix Scientific

056541-1G
101910-842EA 103.5 USD
101910-842
1,3-Difluoroacetone ≥98%
1,3-Difluoroacetone

Hazard: Flammable/Toxic/Refrigerate

Formula: C₃H₄F₂O
Boiling Pt: 118…120 °C
Density: 1.3066
Flash Pt: 51.1 °C
MDL Number: MFCD07368665
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR