4-(Trifluoromethyl)pyridine-2-sulfonic acid amide ≥97%

Supplier: Matrix Scientific

056540-500MG
101910-840EA 315.38 USD
101910-840
4-(Trifluoromethyl)pyridine-2-sulfonic acid amide ≥97%
4-(Trifluoromethyl)pyridine-2-sulfonic acid amide

Hazard: Irritant

Formula: C₆H₅F₃N₂O₂S
MDL Number: MFCD16875529
CAS Number: 1252046-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR