4-Chloro-2-methyl-1,8-naphthyridine ≥95%

Supplier: Matrix Scientific

056538-1G
101910-836EA 320 USD
101910-836
4-Chloro-2-methyl-1,8-naphthyridine ≥95%
4-Chloro-2-methyl-1,8-naphthyridine

Hazard: Irritant

Formula: C₉H₇ClN₂
MDL Number: MFCD16140201
CAS Number: 1221272-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR