3,5-Dibromothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

056531-1G
101910-826EA 75.38 USD
101910-826
3,5-Dibromothiophene-2-carboxylic acid ≥95%
3,5-Dibromothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₅H₂Br₂O₂S
MDL Number: MFCD01006744
CAS Number: 7311-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR