N-[2-(3,4-Dimethoxyphenyl)ethyl]-2-(N'-hydroxycarbamimidoyl)acetamide ≥95%

Supplier: Matrix Scientific

056481-1G 056481-500MG
101910-672EA 1400 USD
101910-672 101910-670
N-[2-(3,4-Dimethoxyphenyl)ethyl]-2-(N'-hydroxycarbamimidoyl)acetamide ≥95%
N-[2-(3,4-Dimethoxyphenyl)ethyl]-2-(N'-hydroxycarbamimidoyl)acetamide

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₄
MDL Number: MFCD00955467
CAS Number: 1308764-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR