2-(2,2-Dimethoxyethyl)aniline ≥95%

Supplier: Matrix Scientific

056479-500MG 056479-1G
101910-662EA 927.69 USD
101910-662 101910-660
2-(2,2-Dimethoxyethyl)aniline ≥95%
2-(2,2-Dimethoxyethyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD09833913
CAS Number: 150760-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR