4,5,5,5-Tetrafluoro-4-(trifluoromethoxy)-pent-2-enoic acid ≥95%

Supplier: Matrix Scientific

056461-500MG 056461-1G
101910-592EA 467.69 USD
101910-592 101910-590
4,5,5,5-Tetrafluoro-4-(trifluoromethoxy)-pent-2-enoic acid ≥95%
4,5,5,5-Tetrafluoro-4-(trifluoromethoxy)-pent-2-enoic acid

Hazard: Irritant

Formula: C₆H₃F₇O₃
MDL Number: MFCD00155978
CAS Number: 261760-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR