tert-Butyl piperidine-4-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

056459-1G
101910-584EA 29.23 USD
101910-584
tert-Butyl piperidine-4-carboxylate hydrochloride ≥95%
tert-Butyl piperidine-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD03788468
CAS Number: 892493-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR