tert-Butyl-N-[3-(1-aminoethyl)phenyl]carbamate ≥95%

Supplier: Matrix Scientific

056449-500MG
101910-544EA 1109.23 USD
101910-544
tert-Butyl-N-[3-(1-aminoethyl)phenyl]carbamate ≥95%
tert-Butyl-N-[3-(1-aminoethyl)phenyl]carbamate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MW: 236.32 g/mol
MDL Number: MFCD03839828
CAS Number: 886362-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR