1-(Piperidin-4-yl)-1H-1,3-benzodiazole hydrochloride ≥95%

Supplier: Matrix Scientific

056442-500MG
101910-520EA 747.69 USD
101910-520
1-(Piperidin-4-yl)-1H-1,3-benzodiazole hydrochloride ≥95%
1-(Piperidin-4-yl)-1H-1,3-benzodiazole hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃
MDL Number: MFCD02178923
CAS Number: 1187174-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR